Anti-CRYAB Mouse Monoclonal Antibody [clone: S4]

Lieferant: Abnova
ABNOH00001410-M01AEA 380 EUR
ABNOH00001410-M01A
Anti-CRYAB Mouse Monoclonal Antibody [clone: S4]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant CRYAB.

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: S4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR