Anti-ENKUR Mouse Monoclonal Antibody [clone: 4A1]

Lieferant: Abnova
ABNOH00219670-M02EA 394 EUR
ABNOH00219670-M02
Anti-ENKUR Mouse Monoclonal Antibody [clone: 4A1]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant C10orf63.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ENKUR
Clonality: Monoclonal
Clone: 4A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR