Anti-ENKUR Mouse Monoclonal Antibody [clone: 3B3]
Lieferant: Abnova
ABNOH00219670-M01EA 394 EUR
ABNOH00219670-M01
Anti-ENKUR Mouse Monoclonal Antibody [clone: 3B3]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant C10orf63.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: ENKUR
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human