Anti-ARL13B Mouse Monoclonal Antibody [clone: 6F11]
Lieferant: Abnova
ABNOH00200894-M02EA 394 EUR
ABNOH00200894-M02
Anti-ARL13B Mouse Monoclonal Antibody [clone: 6F11]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant ARL13B.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: ARL13B
Clonality: Monoclonal
Clone: 6F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen