Anti-DGKK Mouse Monoclonal Antibody [clone: 4G12]

Lieferant: Abnova
ABNOH00139189-M01EA 394 EUR
ABNOH00139189-M01
Anti-DGKK Mouse Monoclonal Antibody [clone: 4G12]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant DGKK.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: DGKK
Clonality: Monoclonal
Clone: 4G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR