Anti-CCL28 Mouse Monoclonal Antibody [clone: 3A7]

Lieferant: Abnova
ABNOH00056477-M03EA 394 EUR
ABNOH00056477-M03
Anti-CCL28 Mouse Monoclonal Antibody [clone: 3A7]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant CCL28.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CCL28
Clonality: Monoclonal
Clone: 3A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR