Anti-GIPC3 Mouse Monoclonal Antibody [clone: 8H8]

Lieferant: Abnova
ABNOH00126326-M01EA 394 EUR
ABNOH00126326-M01
Anti-GIPC3 Mouse Monoclonal Antibody [clone: 8H8]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant GIPC3.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: GIPC3
Clonality: Monoclonal
Clone: 8H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR