Anti-DSN1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28597EA 545 EUR
ABNOPAB28597
Anti-DSN1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant DSN1.

Recommended Dilutions: Western Blot: 1:100-1:250; Immunofluorescence: 1-4 ?g/ml; Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: DSN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR