Anti-HORMAD2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28648EA 545 EUR
ABNOPAB28648
Anti-HORMAD2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant HORMAD2.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Western Blot: 1:100-1:250

Type: Primary
Antigen: HORMAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR