Anti-RAP1GAP Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28657EA 545 EUR
ABNOPAB28657
Anti-RAP1GAP Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant RAP1GAP.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:100-1:250

Type: Primary
Antigen: RAP1GAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR