Anti-ITPR1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S24-18]

Lieferant: Abnova
ABNOMAB11687EA 545 EUR
ABNOMAB11687
Anti-ITPR1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S24-18]
Antikörper
Mouse monoclonal antibody raised against partial synthetic protein of Itpr1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: ITPR1
Clonality: Monoclonal
Clone: S24-18
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR