Anti-KCNMB4 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S18A-3]

Lieferant: Abnova
ABNOMAB11737EA 545 EUR
ABNOMAB11737
Anti-KCNMB4 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S18A-3]
Antikörper
Mouse monoclonal antibody raised against partial synthetic protein of Kcnmb4 .

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: KCNMB4
Clonality: Monoclonal
Clone: S18A-3
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR