Anti-SLC14A2 Rabbit Polyclonal Antibody (ATTO 488)

Lieferant: Abnova
ABNOPAB28822EA 545 EUR
ABNOPAB28822
Anti-SLC14A2 Rabbit Polyclonal Antibody (ATTO 488)
Antikörper
Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.

Recommended Dilutions: Immunofluorescence Western Blot: 1:1000

Type:
Antigen: SLC14A2
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR