Anti-AQP1 Rabbit Polyclonal Antibody (ATTO 390)

Lieferant: Abnova
ABNOPAB28839EA 545 EUR
ABNOPAB28839
Anti-AQP1 Rabbit Polyclonal Antibody (ATTO 390)
Antikörper
Rabbit polyclonal antibody raised against recombinant Rat Aqp1.

Recommended Dilutions: Immunofluorescence: 1:200-400; Western Blot: 1:1000-4000

Type:
Antigen: AQP1
Clonality: Polyclonal
Clone:
Conjugation: ATTO 390
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR