Anti-TRPV4 Mouse Monoclonal Antibody [clone: 4E11]

Lieferant: Abnova
ABNOH00059341-M01EA 394 EUR
ABNOH00059341-M01
Anti-TRPV4 Mouse Monoclonal Antibody [clone: 4E11]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant TRPV4.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TRPV4
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR