Anti-AQP2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abnova
ABNOPAB28867EA 545 EUR
ABNOPAB28867
Anti-AQP2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Rabbit polyclonal antibody raised against recombinant Rat Aqp2.

Recommended Dilutions: Immunofluorescence: 1:400; Western Blot: 1:1000-4000

Type:
Antigen: AQP2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR