Anti-KCNB1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S4-11]
Lieferant: Abnova
ABNOMAB11809EA 545 EUR
ABNOMAB11809
Anti-KCNB1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S4-11]
Antikörper
Mouse monoclonal antibody raised against partial recombinant Kcnb1.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: KCNB1
Clonality: Monoclonal
Clone: S4-11
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen