Anti-APP Rabbit Polyclonal Antibody (ATTO 390)
Lieferant: Abnova
ABNOPAB28911EA 545 EUR
ABNOPAB28911
Anti-APP Rabbit Polyclonal Antibody (ATTO 390)
Antikörper
Rabbit polyclonal antibody against amyloid fibrils and fibrillar oligomers.
Recommended Dilutions: Dot Blot: 1:1000
Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: ATTO 390
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen