Anti-KCNA6 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S19-36]

Lieferant: Abnova
ABNOMAB11827EA 545 EUR
ABNOMAB11827
Anti-KCNA6 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S19-36]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of Kcna6.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: KCNA6
Clonality: Monoclonal
Clone: S19-36
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR