Anti-KDEL Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: Abnova
ABNOPAB28973EA 545 EUR
ABNOPAB28973
Anti-KDEL Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of KDEL.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000
Type:
Antigen: KDEL
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host:
Isotype:
Reactivity:
Bestellen