Anti-SOD2 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Abnova
ABNOPAB28993EA 545 EUR
ABNOPAB28993
Anti-SOD2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Rabbit polyclonal antibody raised against Recombinant of Sod2.
Recommended Dilutions: Western Blot: 0.5 ?g/ml
Type:
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen