Anti-KRT1 Mouse Monoclonal Antibody [clone: SPM591]

Lieferant: Abnova
ABNOMAB12075EA 440 EUR
ABNOMAB12075
Anti-KRT1 Mouse Monoclonal Antibody [clone: SPM591]
Antikörper
Mouse monoclonalantibody raised against Basic Cytokeratin.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: KRT1
Clonality: Monoclonal
Clone: SPM591
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR