Anti-RAB5 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abnova
ABNOPAB29011EA 492 EUR
ABNOPAB29011
Anti-RAB5 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of RAB5A.

Recommended Dilutions: Immunofluorescence Western Blot: 1:1000

Type:
Antigen: RAB5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR