Anti-SLC9A3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Abnova
ABNOPAB29031EA 545 EUR
ABNOPAB29031
Anti-SLC9A3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of Slc9a3.
Recommended Dilutions: Immunocytochemistry Western Blot: 1:1000
Type:
Antigen: SLC9A3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen