Anti-RCOR1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S72-8]

Lieferant: Abnova
ABNOMAB11935EA 545 EUR
ABNOMAB11935
Anti-RCOR1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: S72-8]
Antikörper
Mouse monoclonal antibody raised against partial recombinant RCOR1.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: RCOR1
Clonality: Monoclonal
Clone: S72-8
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR