Anti-GFAP Chicken Polyclonal Antibody

Lieferant: Abnova
ABNOPAB29075EA 364 EUR
ABNOPAB29075
Anti-GFAP Chicken Polyclonal Antibody
Antikörper
Chicken polyclonal antibody raised against recombinant Rat Gfap.

Recommended Dilutions: Immunocytochemistry: 1:2000-1:5000; Immunohistochemistry: 1:2000-1:5000; Western Blot: 1:5000-1:10000

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR