Anti-MAP2 Chicken Polyclonal Antibody

Lieferant: Abnova
ABNOPAB29077EA 655 EUR
ABNOPAB29077
Anti-MAP2 Chicken Polyclonal Antibody
Antikörper
Chicken polyclonal antibody raised against recombinant Human MAP2.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000; Western Blot: 1:2000-1:5000

Type: Primary
Antigen: MAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR