Anti-Clostridium difficile Toxin B Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Abnova
ABNOPAB29155EA 545 EUR
ABNOPAB29155
Anti-Clostridium difficile Toxin B Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Chicken polyclonal antibody raised against Clostridium difficile Toxin B.

Type:
Antigen: Clostridium difficile Toxin B
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR