Anti-IgY F(c) Monoclonal Antibody (Biotin)

Lieferant: Abnova
ABNOPAB29162EA 229 EUR
ABNOPAB29162
Anti-IgY F(c) Monoclonal Antibody (Biotin)
Antikörper
Donkey anti-chicken IgY recognizes chicken IgY F(c). The anti-F(c)-specific fraction was purified using antigen affinity chromatography.

Type: Secondary
Antigen: IgY F(c)
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope: F(c)
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR