Anti-HBA1 Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Abnova
ABNOPAB29178EA 338 EUR
ABNOPAB29178
Anti-HBA1 Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Chicken polyclonal antibody raised against native Human Hemoglobin with HRP conjugation.

Type:
Antigen: HBA1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR