Anti-HIV1 gp120 Mouse Monoclonal Antibody [clone: 1G12]
Lieferant: Abnova
ABNOMAB0036-M08AEA 344 EUR
ABNOMAB0036-M08A
Anti-HIV1 gp120 Mouse Monoclonal Antibody [clone: 1G12]
Antikörper
Mouse monoclonal antibody raised against human Immunodeficiency Virus type 1 (HIV-1), CN54 gp120 outer domain.
Type: Primary
Antigen: HIV1 gp120
Clonality: Monoclonal
Clone: 1G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen