Anti-SUMO2 Mouse Monoclonal Antibody [clone: SPM572]

Lieferant: Abnova
MAB12055
ABNOMAB12055EA 523 EUR
ABNOMAB12055
Anti-SUMO2 Mouse Monoclonal Antibody [clone: SPM572]
Antikörper
Mouse monoclonal antibody raised against SUMO2/SUMO3.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: SUMO2
Clonality: Monoclonal
Clone: SPM572
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR