Anti-CD46 Mouse Monoclonal Antibody [clone: 122.2]

Lieferant: Abnova
ABNOMAB12137EA 471 EUR
ABNOMAB12137
Anti-CD46 Mouse Monoclonal Antibody [clone: 122.2]
Antikörper
Mouse monoclonal antibody raised against recombinant human CD46.

Recommended Dilutions: Flow Cytometry Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: 122.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR