Anti-DSG1 Mouse Monoclonal Antibody [clone: CA72/733]

Lieferant: Abnova
ABNOMAB12147EA 440 EUR
ABNOMAB12147
Anti-DSG1 Mouse Monoclonal Antibody [clone: CA72/733]
Antikörper
Mouse monoclonal antibody raised against recombinant human DSG1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: DSG1
Clonality: Monoclonal
Clone: CA72/733
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR