Anti-HIF1A Mouse Monoclonal Antibody [clone: 2E23]

Lieferant: Abnova
ABNOH00003091-M04EA 380 EUR
ABNOH00003091-M04
Anti-HIF1A Mouse Monoclonal Antibody [clone: 2E23]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant HIF1A.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone: 2E23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR