Anti-WASF1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S91-36]

Lieferant: Abnova
ABNOMAB11596EA 545 EUR
ABNOMAB11596
Anti-WASF1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S91-36]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of WASF1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: WASF1
Clonality: Monoclonal
Clone: S91-36
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR