Anti-IFIH1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28678EA 545 EUR
ABNOPAB28678
Anti-IFIH1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant IFIH1, domain 1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: IFIH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR