Anti-EMILIN1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28690EA 545 EUR
ABNOPAB28690
Anti-EMILIN1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant EMILIN1.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:100-1:250

Type: Primary
Antigen: EMILIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR