Anti-TROVE2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28693EA 545 EUR
ABNOPAB28693
Anti-TROVE2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant TROVE2.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: TROVE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR