Anti-SDHB Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28696EA 545 EUR
ABNOPAB28696
Anti-SDHB Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant SDHB, subunit B.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: SDHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR