Anti-PHKG2 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00005261-B01PEA 380 EUR
ABNOH00005261-B01P
Anti-PHKG2 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human PHKG2 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: PHKG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen