Anti-OLFM1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S96-7]

Lieferant: Abnova
ABNOMAB11615EA 545 EUR
ABNOMAB11615
Anti-OLFM1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S96-7]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of OLFM1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: OLFM1
Clonality: Monoclonal
Clone: S96-7
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR