Anti-CILP Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28713EA 545 EUR
ABNOPAB28713
Anti-CILP Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant CILP.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: CILP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR