Anti-TAT Mouse Monoclonal Antibody [clone: 5G6]
Lieferant: Abnova
ABNOH00006898-M01EA 394 EUR
ABNOH00006898-M01
Anti-TAT Mouse Monoclonal Antibody [clone: 5G6]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant TAT.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: TAT
Clonality: Monoclonal
Clone: 5G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen