Anti-SCNN1A Rabbit Polyclonal Antibody (ATTO 594)

Lieferant: Abnova
ABNOPAB28770EA 545 EUR
ABNOPAB28770
Anti-SCNN1A Rabbit Polyclonal Antibody (ATTO 594)
Antikörper
Rabbit polyclonal antibody raised against recombinant Rat Scnn1a.

Recommended Dilutions: Immunofluorescence: 1:400; Immunohistochemistry Western Blot: 1:1000

Type:
Antigen: SCNN1A
Clonality: Polyclonal
Clone:
Conjugation: ATTO 594
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR