Anti-WDR62 Mouse Monoclonal Antibody [clone: 3H6]

Lieferant: Abnova
ABNOH00284403-M04EA 394 EUR
ABNOH00284403-M04
Anti-WDR62 Mouse Monoclonal Antibody [clone: 3H6]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant WDR62.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: WDR62
Clonality: Monoclonal
Clone: 3H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR