Anti-RARRES2 Mouse Monoclonal Antibody [clone: 2F6]

Lieferant: Abnova
ABNOH00005919-M19EA 380 EUR
ABNOH00005919-M19
Anti-RARRES2 Mouse Monoclonal Antibody [clone: 2F6]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant RARRES2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: RARRES2
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR