Anti-EFEMP2 Mouse Monoclonal Antibody [clone: 3H7]

Lieferant: Abnova
ABNOH00030008-M36EA 394 EUR
ABNOH00030008-M36
Anti-EFEMP2 Mouse Monoclonal Antibody [clone: 3H7]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant EFEMP2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: EFEMP2
Clonality: Monoclonal
Clone: 3H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR