Anti-DPYSL2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB29239EA 356 EUR
ABNOPAB29239
Anti-DPYSL2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human DPYSL2.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1000

Type: Primary
Antigen: DPYSL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR