Anti-XAGE1D Mouse Monoclonal Antibody [clone: 4D12]

Lieferant: Abnova
ABNOH00009503-M01EA 394 EUR
ABNOH00009503-M01
Anti-XAGE1D Mouse Monoclonal Antibody [clone: 4D12]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant XAGE1D.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: XAGE1D
Clonality: Monoclonal
Clone: 4D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR